Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Baker-flex™, Cellulose PEI-F, J.T.Baker™

Click to view available options
Quantity:
25 sheets
50 sheets

Chemical Identifiers

9004-34-6

Flexible TLC Sheet

Solubility Information Insoluble in water
Physical Form Crystal or Powder
Odor Odorless
Color White
Quantity 50 sheets
Chemical Name or Material Cellulose PEI-F