Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Baker-flex™, Cellulose PEI, J.T. Baker™

Catalog Number 02002562
Click to view available options
Quantity:
25 Sheets
50 Sheets
This item is not returnable. View return policy
9004-34-6
This item is not returnable. View return policy
9004-34-6

Flexible TLC Sheet

Color White
Physical Form Crystal or Powder
Solubility Information Insoluble in water
Quantity 50 Sheets
Odor Odorless
Chemical Name or Material Cellulose PEI
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit