Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Avantor Baker-flex, Silica Gel IB, J.T.Baker
Flexible TLC Sheet
Manufacturer: Avantor 446204
Description
Flexible TLC Sheet
Specifications
Silica Gel IB | |
2230°C | |
White | |
Odorless | |
60.09 |
7631-86-9 | |
1710°C | |
25 Sheets | |
Insoluble aq. soln. | |
Crystal or Powder |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title