Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avantor™ BAKERBOND™ PROchievA™ Recombinant Protein A Resin, Affinity Chromatography Resin

Offers superior dynamic binding capacity with excellent alkaline stability linked to Avantor's proprietary ligand.

$1844.00 - $2765.00

Specifications

Content And Storage 2°C to 8°C
Particle Size 68 to 78 μm
View More Specs

Products 2
Catalog Number Mfr. No. Sample Volume Price Quantity & Availability  
Catalog Number Mfr. No. Sample Volume Price Quantity & Availability  
02-004-669
View Documents
Avantor™
789901
25 mL
Each for $1,844.00
Add to cart
 
02-004-670
View Documents
Avantor™
789902
100 mL
Each for $2,765.00
Add to cart
 
Description

Description

  • Offers superior dynamic binding capacity with excellent alkaline stability linked to Avantor's proprietary ligand
  • The new protein A resin is designed for high-performance in the critical affinity chromatography step used in the purification of mAbs, Fc fusion proteins, and other similar IgG-like molecules
  • Designed and manufactured by Avantor to the high standards established for our J.T. Baker™ brand chemicals
  • BAKERBOND™ PROchievA™ recombinant protein A resin can be used with other J.T. Baker™ BAKERBOND™ resins to enhance the separation power of your chromatography process steps
Specifications

Specifications

2°C to 8°C
68 to 78 μm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit