Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAKERBOND spe™ Aromatic Sulfonic Acid (C6H5SO3H) Disposable Extraction Columns, J.T. Baker™
Click to view available options
Quantity:
30/Box
400/Box
50/Box
Description
Disposable Extraction Columns
Specifications
Odor | Odorless |
Quantity | 50/Box |
Physical Form | Solid |
Solubility Information | Practically Insoluble |
Chemical Name or Material | Aromatic Sulfonic Acid (C6H5SO3H) Disposable Extraction Columns |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction