Learn More
Avantor™ BAKERBOND Speedisk™ Extraction Disks
Maximize capacity, adsorption and SPE speed. Avantor BAKERBOND Speedisk Extraction Disks are neither cartridge nor membrane that provides an excellent SPE tool for environmental samples and combines a thin bed of BAKERBOND sorbent with a special laminar configuration for testing drinking water, ground water, waste water, solid waste or process streams.
Supplier:  Avantor™ 805606
 View more versions of this product
Catalog No. 02-002-377
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Pre-assembled with rigid support and baffle to maximize laminar flow, allow for filtration, resist clogging, and maximize capacity, adsorption, and speed
  • Require minimal glassware, set up time, and assembly
  • Minimizes impact of extractable compounds
  • Resists clogging and ensures high throughput rate
  • Reduce SPE run times
Specifications
Specifications
Dirty Samples: EPA Methods 608,846, and Slightly Polar to Non-Polar Industrial Samples
C18 XF
20/Pk.
Expanded Extraction Station (8095-06)
Octadecyl (C18 XF) Extraction Disk
50 mm
Product Suggestions
Product Suggestions