Learn More
Cell Counters Available on GSA/VA Contract for Federal Government customers only.
For automatic calculation including percentages
Manufacturer:  Tip Industries, Inc. 202C
 View more versions of this product
Catalog No. 02-670-12
  Check Availability
Add to cart
Description
Description
For automatic calculation including percentages
  • Totaling unit rings a bell at 100 increments
  • Dual reset knobs
  • Last unit is the totaling unit, which records total number of strokes made on other keys
  • Removable tabs on top of counter provide area for identification of classification being counted
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
0 to 999
2 Counting units; Single reset knobs
Cell counting with automatic calculations
Differential Cell Counter