Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Balicatib
Potent and selective cathepsin K inhibitor
$235.48 - $882.76
Specifications
Chemical Name or Material | N-[1-(Cyanomethylcarbamoyl)cyclohexyl]-4-(4-propylpiperazin-1-yl)benzamide |
---|---|
CAS | 354813-19-7 |
Target | Synthetic Cathepsin Inhibitors |
Molecular Formula | C23H33N5O2 |
Formula Weight | 411.54 |
Description
Potent and selective cathepsin K inhibitor (IC50 values are 1.4, 56 and 480 nM for human, rat and mouse cathepsin K, respectively). Exhibits >300-fold selectivity for cathepsin K over cathepsins L, B and S. Long term supratherapeutic dosing increases tissue levels of cathepsins L and B in rats.Specifications
N-[1-(Cyanomethylcarbamoyl)cyclohexyl]-4-(4-propylpiperazin-1-yl)benzamide | |
Synthetic Cathepsin Inhibitors | |
411.54 |
354813-19-7 | |
C23H33N5O2 | |
>98% |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title