Learn More
Kemtec™ Ballistics Kits: The Sacco & Vanzetti Case Products for Science Education
Kemtec™ Ballistics Kits: The Sacco & Vanzetti Case allows students to decide whether suspects are guilty or innocent by examining case evidence
$687.00 - $720.00
Specifications
For Use With (Application) Grades 7-12
Product Line Ballistics
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S30808 Products for Science Education
View Documents
Kemtec™
10554
Complete Kit
Each for $720.00
Add to cart
S30809 Products for Science Education
View Documents
Kemtec™
10555
Expansion Kit
Each for $687.00
Add to cart
Description
Description
Complete Kit:
  • Can aspiring ballistics experts determine if Sacco and Vanzetti committed this crime?
  • Reconstruct the crime scene from evidence, then collect evidence from the scene
  • Calculate projectile trajectories using a map of gunshot evidence, ballistics blocks, and trajectory rods
  • Learn to predict bullet trajectories using both string and laser techniques
  • Characterize ammunition using inactive dummy cartridges, fired bullets, and fired casings
  • Use gunshot residue analysis to identify suspects
  • Analyze crime scene evidence to support the court case
  • A great opportunity for teaching chain of custody and proper evidence collection
  • Includes instructions and documents for holding a mock trial
  • Includes supporting cross curricular extension activities to address social and political issues
Expansion Set:
  • Want to run more students per experiment simultaneously avoiding stations?
  • Need to accommodate multiple sessions or growing Forensic Science classroom sizes?
  • Expansion kit provides additional materials for trajectory analysis and bullet measuring experiments
  • Includes:
    • Set of four ballistics blocks with trajectory rods
    • Two lasers with ballistics block adapters
    • Three sets of chemical resistant ballistics evidence maps
    • Six stainless steel calipers
Specifications
Specifications
Grades 7-12
Ballistics