Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Balsam Canada, Natural, Vion Biosciences
Shop All Aldon Products

Click to view available options
Quantity:
25 g
Description
- Use natural balsam canada resin in xylene as a mounting medium to prepare permanent sample slides.
Specifications
Specifications
| Applications | Slide Mounting Media |
| Aqueous or Permanent | Permanent |
| Volume (English) | 30 CC |
| Quantity | 25 g |
| Product Type | Mounting Medium |
| Packaging | Amber Glass |
| Content And Storage | Store in a cool, dry, well-ventilated area away from incompatible substances. Keep away from ignition sources. Protect from light. |
| Product Line | Vion Histology |
| Reagent Type | Mounting Solution |
| Volume (Metric) | 30 mL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction