Learn More
Parker Hannifan BALSTON DFU CQ TUBE 10/PK
SDP
Supplier:  Parker Hannifan 993305CQ
Encompass_Preferred
Tube; General Purpose DFU - Low Flow Gas; Nylon; I.V: 0.01L; Gas Efficiency(at .01um): 98%; Liq. Efficiency(98% retention): 8um; U-Type Filter: Polyolifin binders with borosilicate glass fiber produces pure while filter media
Catalog No. NC9441594
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.