Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAM 7, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
331244-89-4
C21H19N5O2S
405.476
KOCVKGYKBLJEPK-LYBHJNIJSA-N
bam7,e-4-2-2-ethoxyphenyl hydrazono-3-methyl-1-4-phenylthiazol-2-yl-1h-pyrazol-5 4h-one,3-methyl-1-4-phenyl-2-thiazolyl-1h-pyrazole-4,5-dione 4-2-2-ethoxyphenyl hydrazone,bam 7,bax activator molecule 7,4-2-2-ethoxyphenyl hydrazono-3-methyl-1-4-phenylthiazol-2-yl-1h-pyrazol-5 4h-one,4e-4-2-ethoxyphenyl hydrazinylidene-5-methyl-2-4-phenyl-1,3-thiazol-2-yl pyrazol-3-one,4z-4-2-2-ethoxyphenyl hydrazin-1-ylidene-5-methyl-2-4-phenyl-1,3-thiazol-2-yl pyrazol-3-one,e-4-2-2-ethoxyphenyl hydrazono-5-methyl-2-4-phenylthiazol-2-yl-2,4-dihydro-3h-pyrazol-3-one
5754137
(4E)-4-[(2-ethoxyphenyl)hydrazinylidene]-5-methyl-2-(4-phenyl-1,3-thiazol-2-yl)pyrazol-3-one
CCOC1=CC=CC=C1NN=C2C(=NN(C2=O)C3=NC(=CS3)C4=CC=CC=C4)C
This item is not returnable. View return policy
331244-89-4
C21H19N5O2S
405.476
KOCVKGYKBLJEPK-LYBHJNIJSA-N
bam7,e-4-2-2-ethoxyphenyl hydrazono-3-methyl-1-4-phenylthiazol-2-yl-1h-pyrazol-5 4h-one,3-methyl-1-4-phenyl-2-thiazolyl-1h-pyrazole-4,5-dione 4-2-2-ethoxyphenyl hydrazone,bam 7,bax activator molecule 7,4-2-2-ethoxyphenyl hydrazono-3-methyl-1-4-phenylthiazol-2-yl-1h-pyrazol-5 4h-one,4e-4-2-ethoxyphenyl hydrazinylidene-5-methyl-2-4-phenyl-1,3-thiazol-2-yl pyrazol-3-one,4z-4-2-2-ethoxyphenyl hydrazin-1-ylidene-5-methyl-2-4-phenyl-1,3-thiazol-2-yl pyrazol-3-one,e-4-2-2-ethoxyphenyl hydrazono-5-methyl-2-4-phenylthiazol-2-yl-2,4-dihydro-3h-pyrazol-3-one
5754137
(4E)-4-[(2-ethoxyphenyl)hydrazinylidene]-5-methyl-2-(4-phenyl-1,3-thiazol-2-yl)pyrazol-3-one
CCOC1=CC=CC=C1NN=C2C(=NN(C2=O)C3=NC(=CS3)C4=CC=CC=C4)C

Selective Bax activator; induces Bax-mediated apoptosis

Bax activator (EC50 = 3.3 μM); binds at the BH3-binding site. Selective for Bax over other antiapoptotic and proapoptotic proteins. Triggers Bax oligomerization in vitro; induces Bax-mediated apoptosis in mouse embryonic fibroblasts.
Quantity 10mg
Formula Weight 405.47
Percent Purity >99%
Chemical Name or Material BAM 7

Recommended Storage : Store at +4°C