Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ BAN ORL 24

Potent and selective NOP antagonist

Manufacturer:  Tocris™ 3661/50

 View more versions of this product

Catalog No. 36-615-0R


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Potent and selective NOP receptor antagonist (IC50 values are 0.27, 2500, 6700 and > 10000nM for NOP, κ-,µ- and δ-receptors respectively). Inhibits nociceptin-induced stimulation of [35S]-GTPγS binding and Ca2+ mobilization in CHO cells in vitro and antagonizes NOP agonist-induced reduction in locomotor activity in vivo. Brain penetrant.
  • Chemical Name: (2R)-1-(Phenylmethyl)-N-[3-(spiro[isobenzofuran-1(3H),4'-piperidin]-1-yl)propyl-2-pyrrolidinecarboxamide
  • Formula: C27H35N3O2.2HCl
  • CAS Number: 1401463-54-4
  • Solubility: Soluble to 100mM in water and to 100mM in DMSO
  • Purity: >97
Description & Specifications

Specifications

BAN ORL 24
>97
50mg
BAN ORL 24
506.51
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit