Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Band 3 Mouse anti-Human, Clone: Q1/156, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Band 3
Clone Q1/156
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Applications Western Blot, ELISA, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122717
View Documents
Novus Biologicals
NBP30762210UG
100 μg
Each for $352.89
Add to cart
 
NB122718
View Documents
Novus Biologicals
NBP30762220UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

Band 3 Monoclonal antibody specifically detects Band 3 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications

Specifications

Band 3
0.2 mg/ml
Western Blot, ELISA, Immunofluorescence
Purified
RUO
Human
AE 1, AE1MGC126619, Anion exchange protein 1, Anion exchanger 1, band 3 anion transport protein, BND3, CD233, CD233 antigen, DI, EMPB3, EPB3MGC126623, erythrocyte membrane protein band 3, erythroid anion exchange protein, FR, Froese blood group, MGC116750, MGC116753, RTA1A, Solute carrier family 4 member 1, solute carrier family 4, anion exchanger, member 1 (erythrocyte membraneprotein band 3, Diego blood group), solute carrier family 4, anion exchanger, number 1, SW, Swann blood group, Waldner blood group, WD, WD1, WR, Wright blood group
Human fetal liver cells.
Monoclonal
Protein A or G purified
Q1/156
Western Blot 1-2 ug/ml, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Unconjugated
Mouse
Phospho Specific
10 mM PBS with 0.05% BSA
6521
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit