Learn More
band 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 185661AP
Catalog No. 50-173-2180
Add to cart
Description
Description
The Band 3 protein is the most abundant integral protein of human erythrocyte membranes and functions as the major anion transporter polypeptide of erythrocytes. Band 3 is a 90-100 kDa protein, its microheterogeneity is ascribed mainly to the structural heterogeneity of the carbohydrate moiety. The C-terminal portion of the molecule spans the membrane several times, and is responsible for anion transport. The N-terminal portion, comprising almost half of the polypeptide chain of Band 3, is located inside the red cell and interacts with cytoskeletal and cytoplasmic proteins such as ankyrin.
Specifications
Specifications
band 3
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P02730, P04919, P23562
AE 1, AE1, Anion exchange protein 1, Anion exchanger 1, band 3, Band 3 anion transport protein, BND3, CD233, DI, EMPB3, EPB3, FR, RTA1A, SLC4A1, SW, WD, WD1, WR
Rabbit
IgG
150 μL
-20°C
Primary
20533, 24779, 6521
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
SLC4A1
Liquid
SLC4A1
band 3 Fusion Protein Ag13076
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat