Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BANK1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549620

Catalog No. PA549620

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Involved in B-cell receptor (BCR)-induced Ca(2+) mobilization from intracellular stores. Promotes Lyn-mediated phosphorylation of IP3 receptors 1 and 2.
Specifications

Specifications

BANK1
Polyclonal
PBS with 0.09% sodium azide
Q8NDB2
B-cell scaffold protein with ankyrin repeats, BANK1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55024
Flow Cytometry, Western Blot
Unconjugated
BANK1
Liquid
BANK1
KLH conjugated synthetic peptide between 119-146 amino acids from the Central region of human BANK1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit