Tocris™ Banoxantrone dihydrochloride

Prodrug topoisomerase II inhibitor

Manufacturer: Tocris™ 4219/50

 View more versions of this product

Catalog No. 42-195-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors Banoxantrone dihydrochloride
    Purity >98
    Molecular Weight 517.4
    Quantity 50mg
    Product Type Banoxantrone dihydrochloride

    Bioreductive prodrug; activated by hypoxia to yield the topoisomerase II inhibitor AQ4. Displays little to no cytotoxic activity against 60 tumor cell lines tested under oxic conditions (IC50 >100µM). Exhibits anti-tumor efficacy in vivo when combined with oxic cell cytotoxins; enhances the anti-tumor effect of cyclophosphamide (Cat. No. 4091) in a mouse tumor model.

    • Chemical Name: 1,4-Bis[[2-(dimethyloxidoamino)ethyl]amino]-5,8-dihydroxy-9,10-anthracenedione dihydrochloride
    • Formula: C22H28N4O6.2HCl
    • CAS Number: 252979-56-9
    • Solubility: Soluble to 50mM in water and to 25mM in DMSO)
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit