Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAP1 Mouse anti-Human, Clone: BAP1/2665, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30739510UG

 View more versions of this product

Catalog No. NB122263


Add to cart

Description

Description

BAP1 Monoclonal specifically detects BAP1 in Human samples. It is validated for ELISA, Protein Array.
Specifications

Specifications

BAP1
BAP1/2665
Unconjugated
10 mM PBS with 0.05% BSA
BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2
Recombinant human BAP1 protein fragment (around aa 191-326) (exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
ELISA, Peptide Array
0.2 mg/ml
ELISA, Protein Array
Purified
Mouse
IgG Kappa
100 μg
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Tumor Suppressors
Primary
8314
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit