Learn More
BAP1 Mouse anti-Human, Mouse, Clone: C.15200212, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259268
Catalog No. NBP259268
Add to cart
Description
Description
BAP1 Monoclonal antibody specifically detects BAP1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation, Immunofluorescence.
Specifications
Specifications
BAP1
C.15200212
Western Blot 1:1000-1:2000, Immunoprecipitation 1:400, Immunofluorescence 1:1000
Purified
BAP1
The exact immunogen is propietary information.
Protein G purified
RUO
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse
Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
PBS with 0.05% Sodium Azide
BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2
Mouse
IgG1 Kappa
50 ug
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Tumor Suppressors
Primary
8314