Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAP18 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen BAP18
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB126151
View Documents
Novus Biologicals
NBP310625100UL
100 μg
Each for $352.89
Add to cart
 
NB126152
View Documents
Novus Biologicals
NBP31062525UL
25 μg
Each for $106.19
Add to cart
 
Description

Description

BAP18 Polyclonal antibody specifically detects BAP18 in Rat samples. It is validated for Western Blot
Specifications

Specifications

BAP18
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
124944
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Rat
BAP18, BPTF-associated protein of 18 kDa, chromatin complexes subunit BAP18, chromosome 17 open reading frame 49, MGC49942, MLL1/MLL complex subunit C17orf49
The immunogen is a synthetic peptide directed towards the N-terminal region of Rat BAP18 (NP_001120993). Peptide sequence VKRFGDDLNHISCVIKERTVAQIKTTVKRKVYEDSGIPLPAESPKKGPKK
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit