Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAP31 Monoclonal Antibody (1502CT208.31.16)
BAP31 Monoclonal Antibody (1502CT208.31.16)
Supplier: Thermo Scientific MA537677
Description
BAP31 Monoclonal Antibody for Western Blot, IHC (P)

Specifications
BAP31 | |
Monoclonal | |
Unconjugated | |
BCAP31 | |
6C6 antigen; 6C6-AG; 6C6-AG tumor-associated antigen; accessory protein BAP31; B cell receptor associated protein 31; BAP31; Bcap31; B-cell receptor-associated protein 31; BCR-associated protein 31; BCR-associated protein Bap31; CDM; CDM protein; DDCH; DXS1357E; hypothetical protein LOC533949; OTTHUMP00000025978; p28; p28 Bap31; Protein CDM | |
Mouse | |
Protein G | |
RUO | |
10134 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1502CT208.31.16 | |
PBS with 0.09% sodium azide; pH 7.4 | |
P51572 | |
BCAP31 | |
BAP31 recombinant protein. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction