Alfa Aesar™ BAPTA tetraethyl ester, 98+%

Manufacturer: Alfa Aesar™ B2197114

Quantity 25g

 View more versions of this product

Catalog No. AAB2197114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 73630-07-6
    Molecular Formula C30H40N2O10
    Formula Weight 588.66
    MDL Number MFCD00532663
    Synonym 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid tetraethyl ester
    Chemical Name or Material BAPTA tetraethyl ester

    Specifications

    Formula Weight 588.66
    Melting Point 98° to 102°C
    Molecular Formula C30H40N2O10
    MDL Number MFCD00532663
    CAS 73630-07-6
    Synonym 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid tetraethyl ester
    Quantity 25g
    Chemical Name or Material BAPTA tetraethyl ester
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit