Learn More
Cole-Parmer Barb Sanitary Adapter, Mini Tri-Clamp x 3/16" Hose Barb, Polypropylene; 6/pk DFS Item
Manufacturer:  Cole-Parmer 3120038
Masterflex F-TBL-14MPHR075-187PP Barb Sanitary Adapter, Mini Tri-Clamp x 3/16" Hose Barb, Polypropylene; 6/pk, Maintain sanitary conditionsMeet requirements for USP Class VI, FDA, REACH, and California Proposition 65Surface finishes to 15 micron minimizes adhesion or entrapmentPackaged to exacting tolerances under ISO Class 7 cleanroom conditionsCompatible with other Tri-Clamp connections
Catalog No. NC1476950