Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barbituric Acid, 99%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
67-52-7
67-52-7
CAS Max % 1
Percent Purity 0.99
Grade Reagent
Packaging Poly Bottle
Quantity 125 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.