Learn More
Aspen Surgical™ Bard-Parker™ Surgical Blade Handles
For postmortem work, trimming light plaster casts, and similar forceful cutting
Supplier:  Aspen Surgical™ 371060
 View more versions of this product
Catalog No. 08-915B
Add to Cart
Description
Description
For postmortem work, trimming light plaster casts, and similar forceful cutting
  • Strong plastic handles provide secure palm-fitting grip
  • Bard-Parker™ brand
  • Blades slide on smoothly and securely (sold separately)
  • Use blade sizes 10, 11, 12, 15 for handle size 5
  • Use blade sizes 20, 21, 22, 23 and 25 for handle size 6
  • Use blade size 60 for handle size 8
Specifications
Specifications
Plastic
# 10, # 11, # 12, # 15, # 20 Blade for Postmortem Work, Trimming Light Plaster Casts and Similar Forceful Cutting
5.18 in.
Palm Fitting Grip
Non-sterile
Surgical Scalpel Handle
# 6
13.1 cm
No
Product Suggestions
Product Suggestions