Learn More
Aspen Surgical™ Bard-Parker™ Surgical Blades
Offer consistent sharpness, control and strength
Manufacturer:  Aspen Surgical™ 371325
 View more versions of this product
Catalog No. 08-915-1C
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Offer consistent sharpness, control and strength
  • Each blade packaged in foil pouch with liner for maximum protection of blade and user
  • Rib back blade design on select blades offers greater cutting control and strength
  • Available with plastic shield SafetyLock to securely cover the blade edge before and after use
  • Available sterile in carbon and stainless steel and nonsterile in carbon steel
Specifications
Specifications
Non-sterile
Surgical Blade
For tissue separation, and other procedures that require a sharp surgical blade to puncture or cut
#25
Carbon Steel
Rib