Learn More
Selleck Chemical LLC Bardoxolone Methyl
SDP
Supplier:  Selleck Chemical LLC S80781G
Encompass_Preferred
Bardoxolone Methyl (RTA 402 TP-155 NSC 713200 CDDO Methyl Ester CDDO-Me) is an IKK inhibitor showing potent proapoptotic and anti-inflammatory activities Also a potent Nrf2 activator and nuclear factor- B (NF- B) inhibitor Bardoxolone Methyl abrogates ferroptosis Bardoxolone methyl induces apoptosis and autophagy in cancer cells
Catalog No. 50-000-00228