Learn More
Sklar InstrumentsSupplier Diversity Partner Sklar Edge TC Metzenbaum-Nelson Bariatric Dissecting Scissors, Sklar, 9 in., Blunt/Blunt, Curved, Smooth, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Sklar Edge Bariatric Scissors are long dissecting scissors. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 162310
Sklar Edge TC Metzenbaum-Nelson Bariatric Dissecting Scissors, Sklar, 9 in., Blunt/Blunt, Curved, Smooth, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Sklar Edge Tungsten Carbide Metzenbaum-Nelson Bariatric Scissors are long dissecting scissors most commonly used in Bariatric Surgeries such as gastric bypass, gastric banding, and sleeve gastrectomy. They are also used in Nephrectomy, Cystectomy, and Prostatectomy procedures to dissect and cut tissue. Metzenbaum-Nelson scissors tend to be longer than standard Metzenbaum scissors which make them ideal for use in thoracic procedures as well. These scissors are not recommended for cutting sutures or drains. Sklar Edge TC Scissors provide an exceptionally durable and hard cutting edge. These scissors are designed to stay sharp for a significantly longer period of time than standard scissors. Sklar permanently bonds tungsten carbide inserts to the blades of the scissors which maximizes control and precision.
Catalog No. 50-143-7381