Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Barium 1-methoxy-2-propoxide, 25% w/w in 1-methoxy-2-propanol
Click to view available options
Quantity:
5g
25g
100g
Specifications
Concentration or Composition (by Analyte or Components) | 25% w/w in 1-methoxy-2-propanol |
Sensitivity | Air and moisture sensitive |
Density | 1.04 g/mL |
Flash Point | 35°C (95°F) |
Quantity | 100g |
Linear Formula | Ba[OCH(CH3)CH2OCH3]2 |
UN Number | UN2924 |
Formula Weight | 315.55 |
Physical Form | Liquid |
Chemical Name or Material | Barium 1-methoxy-2-propoxide |
Safety and Handling
GHS H Statement
H314-H318-H226-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures