Alfa Aesar™ Barium 1-methoxy-2-propoxide, 25% w/w in 1-methoxy-2-propanol

Manufacturer: Alfa Aesar™ 4125706

Quantity 5g

 View more versions of this product

Catalog No. AA4125706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 130812-47-4
    Linear Formula Ba[OCH(CH3)CH2OCH3]2
    Formula Weight 315.55
    MDL Number MFCD00078021
    UN Number UN2924
    Chemical Name or Material Barium 1-methoxy-2-propoxide

    Specifications

    Formula Weight 315.55
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P302+P352
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    Physical Form Liquid
    UN Number UN2924
    Health Hazard 1 Danger
    Sensitivity Air & Moisture Sensitive
    Concentration 25% w/w in 1-methoxy-2-propanol
    Flash Point 35° (95°F)
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Linear Formula Ba[OCH(CH3)CH2OCH3]2
    MDL Number MFCD00078021
    CAS 130812-47-4
    Density 1.04g/mL
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H314-H318-H226-H302-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful if inhaled.
    Chemical Name or Material Barium 1-methoxy-2-propoxide

    GHS H Statement
    H314-H318-H226-H302-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P302+P352
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit