Alfa Aesar™ Barium 2,4-pentanedionate hydrate

Manufacturer: Alfa Aesar™ 1461322

Quantity 100g

 View more versions of this product

Catalog No. AA1461322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12084-29-6
    Formula Weight 335.56 (Anhydrous)
    MDL Number MFCD00013484
    UN Number UN1564
    Synonym Barium acetylacetonate
    Chemical Name or Material Barium 2,4-pentanedionate hydrate

    Specifications

    Formula Weight 335.56 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    Physical Form Powder
    CAS 12084-29-6
    UN Number UN1564
    Synonym Barium acetylacetonate
    Melting Point >320°C
    Quantity 100g
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    MDL Number MFCD00013484
    Chemical Name or Material Barium 2,4-pentanedionate hydrate

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit