Alfa Aesar™ Barium 2-ethylhexanoate in 2-ethylhexanoic acid, Ba »17.5%

Manufacturer: Alfa Aesar™ 4484706

Quantity 5g

 View more versions of this product

Catalog No. AA4484706

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 2457-01-4
    Molecular Formula C16H30BaO4
    Formula Weight 423.76
    MDL Number MFCD00058696
    Chemical Name or Material Barium 2-ethylhexanoate in 2-ethylhexanoic acid

    Specifications

    Physical Form Viscous Liquid
    Formula Weight 423.76
    Health Hazard 3 GHS P Statement
    P261-P281-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    Concentration Ba ≈17.5%
    Quantity 5g
    Molecular Formula C16H30BaO4
    Health Hazard 2 GHS H Statement
    H331-H361-H302
    Toxic if inhaled.
    Suspected of damaging fertility or the unborn child.
    Harmful if swallowed.
    TSCA TSCA
    MDL Number MFCD00058696
    CAS 2457-01-4
    Color Yellow
    Chemical Name or Material Barium 2-ethylhexanoate in 2-ethylhexanoic acid

    GHS H Statement
    H331-H361-H302
    Toxic if inhaled.
    Suspected of damaging fertility or the unborn child.
    Harmful if swallowed.

    GHS P Statement
    P261-P281-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit