Alfa Aesar™ Barium 2-methoxyethoxide, 99.5% (metals basis), 10% w/v in 2-methoxyethanol

Manufacturer: Alfa Aesar™ 41256-AC

Quantity 25mL

 View more versions of this product

Catalog No. AA41256AC

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 115503-13-4
    Linear Formula Ba(OCH2CH2OCH3)2
    Formula Weight 287.5
    MDL Number MFCD00673672
    UN Number UN3274
    Chemical Name or Material Barium 2-methoxyethoxide

    Specifications

    Formula Weight 287.5
    UN Number UN3274
    Health Hazard 2 GHS H Statement
    H360-H226-H302-H312-H332
    May damage fertility or the unborn child.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Health Hazard 3 GHS P Statement
    P201-P308+P313
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Physical Form Liquid
    Linear Formula Ba(OCH2CH2OCH3)2
    MDL Number MFCD00673672
    Sensitivity Moisture Sensitive
    Health Hazard 1 Danger
    Concentration 10% w/v in 2-methoxyethanol
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER
    CAS 115503-13-4
    Color Yellow
    Quantity 25mL
    Chemical Name or Material Barium 2-methoxyethoxide
    Assay 99.5%, (metals basis)

    GHS H Statement
    H360-H226-H302-H312-H332
    May damage fertility or the unborn child.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P201-P308+P313
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit