Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Barium 2-methoxyethoxide, 99.5% (metals basis), 10% w/v in 2-methoxyethanol

Manufacturer:  Alfa Aesar™ 41256-AE

Quantity 100mL

 View more versions of this product

Catalog No. AA41256AE

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

115503-13-4
287.5
UN3274
Ba(OCH2CH2OCH3)2
MFCD00673672
Barium 2-methoxyethoxide

Specifications

115503-13-4
Ba(OCH2CH2OCH3)2
MFCD00673672
UN3274
10% w/v in 2-methoxyethanol
Barium 2-methoxyethoxide
Liquid
287.5
Moisture Sensitive
Yellow
100mL
99.5%, (metals basis)

Safety and Handling

GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit