Alfa Aesar™ Barium acetate, 99%

Manufacturer: Alfa Aesar™ A103160B

Quantity 1000g

 View more versions of this product

Catalog No. AAA103160B

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 543-80-6
    Molecular Formula Ba(OOCCH3)2
    Formula Weight 255.43
    MDL Number MFCD00012447
    UN Number UN1564
    Merck Index 14,965
    Synonym Acetic acid barium salt
    Chemical Name or Material Barium acetate

    Specifications

    Formula Weight 255.43
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    MDL Number MFCD00012447
    CAS 543-80-6
    Health Hazard 1 Warning
    Quantity 1000g
    Molecular Formula Ba(OOCCH3)2
    UN Number UN1564
    Merck Index 14,965
    Synonym Acetic acid barium salt
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    Chemical Name or Material Barium acetate
    Assay 99%

    Description

    Applications
    It has acted as cationization agent for sensitive multiple reaction monitoring of plasma free fatty acid profiling in a fish oil human intervention. Barium acetate has been used in the process of titrating sulfates of quinine and quinidine in nonaqueous conditions. Electrophoresis in the presence of barium acetate has analytically and preparatively separate glycosaminoglycans. It has been documented to induce severe cardiac disrhythmia from ingestion. It also finds its application as a chemical reagent, textile mordant, catalyst for organic reactions, and paint and varnish driers.

    Solubility
    Soluble in water at 20°C 588g/L. Insoluble in alcohol.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit