Learn More
Barium acetate, ACS, 99.0-102.0%, Thermo Scientific Chemicals
Description
Barium acetate is used as a chemical reagent, textile mordant and lubricant. It is used to prepare other acetates and barium carbonate. It acts as a mordant for printing textile fabrics. It is utilized for drying paints and varnishes. It serves as a catalyst in organic synthesis. It plays an important role in the separation of glycosaminoglycans. Further, it acts as a cationization agent for sensitive multiple reaction monitoring of plasma free fatty acid profiling in a fish oil human intervention. In addition, it is used in the process of titrating sulfates of quinine and quinidine in nonaqueous conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Crystalline |
Quantity | 5 kg |
UN Number | UN1564 |
Merck Index | 14,965 |
Solubility Information | Soluble in water. Insoluble in alcohol. |
Formula Weight | 255.43 |
Percent Purity | 99.0 to 102.0% |
Grade | ACS |
Odor | Odorless |
Density | 2.47 g/mL |
Show More |
Safety and Handling
Hazard Category | H302+H332 |
Hazard Statement | GHS H Statement H302-H332 Harmful if swallowed. Harmful if inhaled. |
Precautionary Statement | P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S. |
EINECSNumber | 208-849-0 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.