Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Acetate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

543-80-6
C4H6BaO4
255.415
ITHZDDVSAWDQPZ-UHFFFAOYSA-L
10980

High quality chemicals for laboratory and specialized industrial use

SMILES CC(=O)[O-].CC(=O)[O-].[Ba+2]
IUPAC Name barium(2+);diacetate
Formula Weight 255.43
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.47kg
Quantity 2.5kg
Chemical Name or Material Barium Acetate
Grade Baker Analyzed™ ACS Reagent