Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Acetate, Granular, ACS, 99.0-102.0%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
1 kg
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
543-80-6
543-80-6
Grade ACS
CAS Max % 1
Percent Purity 99 to 102%
Calcium (Ca) 0.0005
Chloride 1e-05
Heavy Metals 5 ppm
Insoluble Matter 0.0001
Iron (Fe) 1e-05
Packaging Poly Bottle
Potassium (K) 3e-05
Sodium (Na) 5e-05
Strontium (Sr) 0.002
Quantity 1 kg
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.