Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Barium acetate, Puriss. p.a., ACS Reagent, ≥99%, Honeywell Fluka™
Click to view available options
Quantity:
250g
1kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Linear Formula | (CH3COO)2Ba |
UN Number | NONH for all modes of transport |
Beilstein | 3693411 |
Formula Weight | 255.42g/mol |
Percent Purity | ≥99% |
Grade | ACS/puriss. p.a. |
Quantity | 250g |
Chemical Name or Material | Barium Acetate |
Safety and Handling
P261-P301 + P312 + P330
H302 + H332
EINECSNumber : 208-849-0