Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Barium acetate, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1112606

Quantity 5g

 View more versions of this product

Catalog No. AA1112606

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

543-80-6
255.43
UN1564
14,965
Barium acetate
Ba(OOCCH3)2
MFCD00012447
3693411
Acetic acid barium salt
Puratronic™

Specifications

543-80-6
Ba(OOCCH3)2
MFCD00012447
3693411
Acetic acid barium salt
Odorless
5g
99.999%, (metals basis)
Crystalline
255.43
UN1564
14,965
2.47g/mL
White
Barium acetate
Puratronic™

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

RTECSNumber : AF4550000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit