Learn More
Barium aluminum oxide, tech., Thermo Scientific Chemicals
Description
Barium aluminum oxide is used for getter alloys in vacuum tubes. It also finds application as deoxidizer for copper, Frary’s metal, lubricant for anode rotors in x-ray tubes and spark-plug alloys. It is used to prepare heat-resistant refractory and super-refractory concretes. Further, it gives a better protection against gamma radiation and X-rays.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 100 g |
UN Number | UN1564 |
Solubility Information | Insoluble in water. |
Formula Weight | 561.98 |
Grade | Technical |
Odor | Odorless |
Chemical Name or Material | Barium aluminum oxide |
Safety and Handling
Hazard Category | H302+H332 |
Hazard Statement | GHS H Statement H302-H332 Harmful if swallowed. Harmful if inhaled. |
Precautionary Statement | P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S. |
EINECSNumber | 234-445-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.