Alfa Aesar™ Barium bis(trifluoromethylsulfonyl)imide

Manufacturer: Alfa Aesar™ 46381MD

Quantity 250mg

 View more versions of this product

Catalog No. AA46381MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 168106-22-7
    Molecular Formula C4BaF12N2O8S4
    Linear Formula [(CF3SO2)2N]2Ba
    Formula Weight 697.6
    MDL Number MFCD22200529
    UN Number UN1759
    Synonym Barium triflimide; Barium bis(trifluoromethanesulfonimide)
    Chemical Name or Material Barium bis (trifluoromethylsulfonyl)imide

    Specifications

    Formula Weight 697.6
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Solid
    Linear Formula [(CF3SO2)2N]2Ba
    MDL Number MFCD22200529
    Synonym Barium triflimide; Barium bis(trifluoromethanesulfonimide)
    Melting Point >300°C
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
    Molecular Formula C4BaF12N2O8S4
    Sensitivity Moisture Sensitive
    CAS 168106-22-7
    UN Number UN1759
    Color White to pale yellow
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 1 Danger
    Quantity 250mg
    Chemical Name or Material Barium bis (trifluoromethylsulfonyl)imide

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit