Alfa Aesar™ Barium bromide, anhydrous, 99%

Manufacturer: Alfa Aesar™ 1679230

Quantity 250g

 View more versions of this product

Catalog No. AA1679230

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10553-31-8
    Formula Weight 297.14
    MDL Number MFCD00003444
    UN Number UN1564
    Merck Index 14,968
    Chemical Name or Material Barium bromide, Anhydrous

    Specifications

    Formula Weight 297.14
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    UN Number UN1564
    Color White
    Quantity 250g
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    Sensitivity Hygroscopic
    MDL Number MFCD00003444
    CAS 10553-31-8
    Merck Index 14,968
    Melting Point 850°C
    Solubility Information Soluble in water
    Physical Form Powder
    Density 4.781g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    TSCA TSCA
    Health Hazard 1 Warning
    Chemical Name or Material Barium bromide, Anhydrous
    Assay 99%

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit