Alfa Aesar™ Barium bromide dihydrate, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1112506

Quantity 5g

 View more versions of this product

Catalog No. AA1112506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7791-28-8
    Formula Weight 333.17 (297.14 Anhydrous)
    MDL Number MFCD00149153
    UN Number UN1564
    Merck Index 14,968
    Chemical Name or Material Barium bromide dihydrate
    Grade Puratronic™

    Specifications

    Formula Weight 333.17 (297.14 Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00149153
    Solubility Information Very soluble in water, soluble in methanol
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    TSCA TSCA
    Physical Form Powder
    Quantity 5g
    Density 3.58g/mL
    UN Number UN1564
    Merck Index 14,968
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    Odor Odorless
    CAS 7791-28-8
    Color White
    Chemical Name or Material Barium bromide dihydrate
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit