Alfa Aesar™ Barium bromide, ultra dry, 99.998% (metals basis)

Manufacturer: Alfa Aesar™ 3567814

Quantity 25g

 View more versions of this product

Catalog No. AA3567814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10553-31-8
    Formula Weight 297.14
    MDL Number MFCD00003444
    UN Number UN1564
    Merck Index 14,968
    Chemical Name or Material Barium bromide, Ultra dry

    Specifications

    Formula Weight 297.14
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN1564
    Merck Index 14,968
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    Physical Form Beads
    Sensitivity Hygroscopic
    Health Hazard 1 Warning
    Density 4.781g/mL
    TSCA TSCA
    Mesh Size −10 Mesh
    Odor Odorless
    MDL Number MFCD00003444
    CAS 10553-31-8
    Color White
    Melting Point 850°C
    Quantity 25g
    Storage Note 1
    • Ampouled under argon
    Chemical Name or Material Barium bromide, Ultra dry
    Assay 99.998%, (metals basis)

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit