Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Carbonate, ACS, 99.0-101.0%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
513-77-9
513-77-9
Grade ACS
CAS Max % 1
Percent Purity 99 to 101%
Calcium (Ca) 0.0005
Chloride 2e-05
Heavy Metals 1e-05
Iron (Fe) 2e-05
Packaging Poly Bottle
Potassium (K) 5e-05
Sodium (Na) 0.0002
Strontium (Sr) 0.007
Sulfide 1e-05
Quantity 500 g
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.