Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Barium Carbonate, Honeywell Fluka™

Click to view available options
Quantity:
50g
250g
1kg
513-77-9
CBaO3
197.335
MFCD00003448
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium carbonate,carbonic acid, barium salt 1:1,barium carbonate 1:1,barium monocarbonate,carbonic acid, barium salt,pigment white 10,caswell no. 069,witherite,ci pigment white 10,bw-p
10563
barium(2+);carbonate
C(=O)([O-])[O-].[Ba+2]
513-77-9
CBaO3
197.335
MFCD00003448
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium carbonate,carbonic acid, barium salt 1:1,barium carbonate 1:1,barium monocarbonate,carbonic acid, barium salt,pigment white 10,caswell no. 069,witherite,ci pigment white 10,bw-p
10563
barium(2+);carbonate
C(=O)([O-])[O-].[Ba+2]

Puriss. p.a., ACS reagent

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Linear Formula BaCO3
UN Number NONH for all modes of transport
Beilstein 7045119
Formula Weight 197.34g/mol
Grade ACS/puriss. p.a.
Quantity 250g
Chemical Name or Material Barium Carbonate

P301 + P312 + P330

H302

EINECSNumber : 208-167-3

RTECSNumber : CQ8600000