Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium carbonate, tech., Alfa Aesar™

Used in the manufacture of glass, brick, clay products, barium chemicals, barium ferrites, and production of photographic papers

Manufacturer:  Alfa Aesar 4347822

Quantity 100g

 View more versions of this product

Catalog No. AA4347822


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

513-77-9
197.35
14,969
Technical Grade
BaCO3
MFCD00003448
Barium carbonate

Specifications

513-77-9
BaCO3
MFCD00003448
Almost insoluble in water. Soluble in NH4Cl solution, dilute HCl, HNO3, acetic acid. Insoluble in most alkalies
Odorless
100g
Technical Grade
Powder
197.35
14,969
4.43
811°C
Barium carbonate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302;
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a;
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CQ8600000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit