Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Barium chlorate monohydrate, 98%

Manufacturer:  Alfa Aesar™ A10128A9

Quantity 10kg

 View more versions of this product

Catalog No. AAA10128A9


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10294-38-9
322.26 (304.24 Anhydrous)
UN1445
Ba(ClO3)2·H2O
MFCD00149147
Barium chlorate monohydrate

Specifications

10294-38-9
322.26 (304.24 Anhydrous)
UN1445
Barium chlorate monohydrate
Ba(ClO3)2·H2O
MFCD00149147
10kg
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H271-H301-H331
May cause fire or explosion
strong oxidizer.
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement
P221-P283-P210-P301+P310-P405-P501a
Take any precaution to avoid mixing with combustibles.
Wear fire/flame resistant/retardant clothing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: BARIUM CHLORATE, SOLID

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit