Alfa Aesar™ Barium chlorate monohydrate, 98%

Manufacturer: Alfa Aesar™ A101280E

Quantity 2500g

 View more versions of this product

Catalog No. AAA101280E

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10294-38-9
    Molecular Formula Ba(ClO3)2·H2O
    Formula Weight 322.26 (304.24 Anhydrous)
    MDL Number MFCD00149147
    UN Number UN1445
    Chemical Name or Material Barium chlorate monohydrate

    Specifications

    Formula Weight 322.26 (304.24 Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P283-P210-P301+P310-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 10294-38-9
    Quantity 2500g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H271-H301-H331
    May cause fire or explosion
    strong oxidizer.
    Toxic if swallowed.
    Toxic if inhaled.
    TSCA TSCA
    Molecular Formula Ba(ClO3)2·H2O
    MDL Number MFCD00149147
    UN Number UN1445
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: BARIUM CHLORATE, SOLID
    Chemical Name or Material Barium chlorate monohydrate
    Assay 98%

    GHS H Statement
    H271-H301-H331
    May cause fire or explosion
    strong oxidizer.
    Toxic if swallowed.
    Toxic if inhaled.

    GHS P Statement
    P221-P283-P210-P301+P310-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit